The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159920.1 | NP_001153392.1 | 9 - 10 | 1559 | 125 |
NM_001160030.1 | NP_001153502.1 | 9 - 10 | 1559 | 125 | |
NM_001160031.1 | NP_001153503.1 | 9 - 10 | 1559 | 125 | |
NM_002019.4 | NP_002010.2 | 9 - 10 | 1559 | 125 | |
XM_011535014.1 | XP_011533316.1 | 9 - 10 | 1559 | 125 | |
XM_017020485.1 | XP_016875974.1 | 9 - 10 | 1559 | 125 | |
GenBank mRNA | AF063657.2 | - | 9 - 10 | 1274 | 125 |
AK292936.1 | - | 9 - 10 | 1559 | 125 | |
AK300392.1 | - | 9 - 10 | 1559 | 125 | |
AK309901.1 | - | 9 - 10 | 1559 | 125 | |
BC039007.1 | - | 9 - 10 | 1510 | 125 | |
BX504049.1 | - | 2 - 3 | 198 | 125 | |
EU360600.1 | - | 9 - 10 | 1559 | 125 | |
EU368830.1 | - | 9 - 10 | 1274 | 125 | |
EU826561.1 | - | 9 - 10 | 1274 | 125 | |
U01134.1 | - | 9 - 10 | 1523 | 125 |