Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NTNG2
- Entrez Gene ID
-
84628
- Gene Name
- netrin G2
- Gene Aliases
-
LHLL9381, Lmnt2, NTNG1, bA479K20.1
- Chromosome Location
-
Chr.9: 132161234 - 132244534 on Build GRCh38
View Full Product Details