Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZBTB20
- Entrez Gene ID
-
26137
- Gene Name
- zinc finger and BTB domain containing 20
- Gene Aliases
-
DPZF, HOF, ODA-8S, PRIMS, ZNF288
- Chromosome Location
-
Chr.3: 114314500 - 115147280 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG025240.1 |
- |
1 |
244 |
|
Chr. 3 - 114334968 - 114335800 [-] on Build GRCh38 |
112 |
GenBankmRNA |
BG402430.1 |
- |
|
83 |
|
Chr. - - [-] on Build |
112 |
GenBankmRNA |
BX487283.1 |
- |
1 |
273 |
|
Chr. 3 - 114335337 - 114335829 [-] on Build GRCh38 |
111 |
GenBankmRNA |
DN991835.1 |
- |
1 |
508 |
|
Chr. 3 - 114335476 - 114336057 [-] on Build GRCh38 |
111 |
GenBankmRNA |
AA056060.1 |
- |
1 |
207 |
|
Chr. 3 - 114335158 - 114335763 [-] on Build GRCh38 |
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 568 |
MIR568 |
MIRN568, hsa-mir-568 |
|
Sense |
Chr. - 114316475 - 114316569 on Build GRCh38 |
Downstream |
ZBTB20 antisense RNA 1 |
ZBTB20-AS1 |
|
12.4kb
|
Anti-sense |
Chr. - 114351811 - 114388978 on Build GRCh38 |
View Full Product Details