The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000476.2 | NP_000467.1 | 6 - 7 | 672 | 109 |
NM_001318121.1 | NP_001305050.1 | 6 - 7 | 659 | 109 | |
NM_001318122.1 | NP_001305051.1 | 5 - 6 | 667 | 109 | |
XM_017014427.1 | XP_016869916.1 | 6 - 7 | 2115 | 109 | |
XM_017014428.1 | XP_016869917.1 | 7 - 8 | 1019 | 109 | |
GenBank mRNA | AB021871.1 | - | 4 - 5 | 519 | 109 |
AK223501.1 | - | 5 - 6 | 547 | 109 | |
AK313758.1 | - | 6 - 7 | 603 | 109 | |
BC001116.2 | - | 6 - 7 | 590 | 109 | |
BC100022.1 | - | 6 - 7 | 619 | 109 | |
BT019580.1 | - | 4 - 5 | 519 | 109 | |
CR541977.1 | - | 4 - 5 | 519 | 109 | |
CR975542.1 | - | 6 - 7 | 659 | 109 | |
EU794628.1 | - | 6 - 7 | 672 | 109 |