The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256910.1 | NP_001243839.1 | 2 - 3 | 747 | 125 |
NM_004728.3 | NP_004719.2 | 2 - 3 | 628 | 125 | |
XM_011540336.1 | XP_011538638.1 | 2 - 3 | 620 | 125 | |
XM_017016910.1 | XP_016872399.1 | 2 - 3 | 538 | 125 | |
GenBank mRNA | AK315585.1 | - | 2 - 3 | 577 | 125 |
BC008071.2 | - | 2 - 3 | 566 | 125 | |
CR749598.1 | - | 2 - 3 | 748 | 125 | |
U41387.1 | - | 2 - 3 | 579 | 125 |