Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMO3
Entrez Gene ID
2328
Gene Name
flavin containing monooxygenase 3
Gene Aliases
FMOII, TMAU, dJ127D3.1
Chromosome Location
Chr.1: 171090873 - 171117819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002294.2 NP_001002294.1 7 - 8 1298 116
NM_001319173.1 NP_001306102.1 8 - 9 1408 116
NM_001319174.1 NP_001306103.1 6 - 7 1109 116
NM_006894.5 NP_008825.4 7 - 8 1281 116
XM_011509345.2 XP_011507647.1 8 - 9 1429 116
GenBank mRNA AK223166.1 - 7 - 8 1290 116
AK296105.1 - 6 - 7 1083 116
AK298406.1 - 8 - 9 1393 116
AK313197.1 - 7 - 8 1271 116
BC032016.1 - 7 - 8 1233 116
KU178077.1 - 6 - 7 1187 116
KU178078.1 - 3 - 4 297 116
Z47552.1 - 7 - 8 1280 116

View Full Product Details