The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323608.1 | NP_001310537.1 | 16 - 17 | 2652 | 81 |
NM_001323609.1 | NP_001310538.1 | 16 - 17 | 2652 | 81 | |
NM_001323611.1 | NP_001310540.1 | 16 - 17 | 2401 | 81 | |
NM_005611.3 | NP_005602.3 | 16 - 17 | 2633 | 81 | |
XM_011523253.2 | XP_011521555.1 | 14 - 15 | 2026 | 81 | |
XM_017023513.1 | XP_016879002.1 | 13 - 14 | 2190 | 81 | |
GenBank mRNA | AK307881.1 | - | 16 - 17 | 2610 | 81 |
BC034490.1 | - | 16 - 17 | 2623 | 81 | |
BX537767.1 | - | 15 - 16 | 2214 | 81 | |
S67171.1 | - | 16 - 17 | 2543 | 81 | |
X74594.1 | - | 16 - 17 | 2344 | 81 | |
X76061.1 | - | 16 - 17 | 2584 | 81 |