Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSHR
Entrez Gene ID
7253
Gene Name
thyroid stimulating hormone receptor
Gene Aliases
CHNG1, LGR3, hTSHR-I
Chromosome Location
Chr.14: 80954989 - 81146302 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005268037.4 XP_005268094.1 1 - 2 224 90
XM_005268039.1 XP_005268096.1 1 - 2 224 90
XM_006720245.1 XP_006720308.1 1 - 2 224 90

View Full Product Details