Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AK7
Entrez Gene ID
122481
Gene Name
adenylate kinase 7
Gene Aliases
AK 7
Chromosome Location
Chr.14: 96392111 - 96489427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152327.3 NP_689540.2 4 - 5 543 154
XM_006720021.2 XP_006720084.1 4 - 5 543 154
XM_006720022.2 XP_006720085.1 4 - 5 543 154
XM_011536403.2 XP_011534705.1 4 - 5 543 154
XM_017020956.1 XP_016876445.1 4 - 5 543 154
XM_017020957.1 XP_016876446.1 4 - 5 543 154
XM_017020958.1 XP_016876447.1 4 - 5 543 154
XM_017020959.1 XP_016876448.1 4 - 5 543 154
XM_017020960.1 XP_016876449.1 4 - 5 543 154
GenBank mRNA AK057426.1 - 4 - 5 543 154
BC035256.1 - 4 - 5 532 154

View Full Product Details