The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152327.3 | NP_689540.2 | 5 - 6 | 656 | 140 |
XM_006720021.2 | XP_006720084.1 | 5 - 6 | 656 | 140 | |
XM_006720022.2 | XP_006720085.1 | 5 - 6 | 656 | 140 | |
XM_011536403.2 | XP_011534705.1 | 5 - 6 | 656 | 140 | |
XM_017020956.1 | XP_016876445.1 | 5 - 6 | 656 | 140 | |
XM_017020957.1 | XP_016876446.1 | 5 - 6 | 656 | 140 | |
XM_017020958.1 | XP_016876447.1 | 5 - 6 | 656 | 140 | |
XM_017020959.1 | XP_016876448.1 | 5 - 6 | 656 | 140 | |
XM_017020960.1 | XP_016876449.1 | 5 - 6 | 656 | 140 | |
GenBank mRNA | AK057426.1 | - | 5 - 6 | 656 | 140 |
BC035256.1 | - | 5 - 6 | 645 | 140 |