The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005715.2 | NP_005706.1 | 1 - 2 | 541 | 100 |
XM_011535378.2 | XP_011533680.1 | 1 - 2 | 1232 | 100 | |
XM_017010152.1 | XP_016865641.1 | 1 - 2 | 1785 | 100 | |
GenBank mRNA | AB020316.1 | - | 1 - 2 | 348 | 100 |
AK292922.1 | - | 1 - 2 | 289 | 100 | |
AK315320.1 | - | 1 - 2 | 249 | 100 | |
BC093668.1 | - | 1 - 2 | 293 | 100 | |
BC093694.1 | - | 1 - 2 | 293 | 100 |