Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 27 - 28 5120 91
NM_080645.2 NP_542376.2 12 - 13 1628 91
XM_011535434.1 XP_011533736.1 27 - 28 5264 91
XM_011535435.1 XP_011533737.1 26 - 27 4991 91
XM_011535436.2 XP_011533738.1 12 - 13 1983 91
XM_017010252.1 XP_016865741.1 26 - 27 5516 91
GenBank mRNA BC146868.1 - 12 - 13 1572 91
U73778.1 - 27 - 28 5067 91

View Full Product Details