Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 64 - 65 9179 98
NM_080645.2 NP_542376.2 49 - 50 5687 98
XM_011535434.1 XP_011533736.1 64 - 65 9323 98
XM_011535435.1 XP_011533737.1 63 - 64 9050 98
XM_011535436.2 XP_011533738.1 49 - 50 6042 98
XM_017010252.1 XP_016865741.1 63 - 64 9575 98
GenBank mRNA AJ420501.1 - 9 - 10 667 98
U73778.1 - 64 - 65 98

View Full Product Details