The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178498.3 | NP_848593.2 | 14 - 15 | 2022 | 145 |
XM_006718155.3 | XP_006718218.1 | 14 - 15 | 1925 | 145 | |
XM_011519920.2 | XP_011518222.1 | 15 - 16 | 1502 | 145 | |
XM_017017244.1 | XP_016872733.1 | 15 - 16 | 1948 | 145 | |
GenBank mRNA | AY299482.1 | - | 14 - 15 | 1731 | 145 |
BC041454.1 | - | 14 - 15 | 1495 | 145 | |
CR747563.1 | - | 3 - 4 | 407 | 145 |