Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC5A12
Entrez Gene ID
159963
Gene Name
solute carrier family 5 member 12
Gene Aliases
SMCT2
Chromosome Location
Chr.11: 26667019 - 26723356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011519920.2 XP_011518222.1 2 - 3 133 71

View Full Product Details