Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJB8
Entrez Gene ID
165721
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
Gene Aliases
CT156, DJ6
Chromosome Location
Chr.3: 128462432 - 128467248 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153330.4 NP_699161.1 3 - 3 2416 71
XM_006713519.3 XP_006713582.1 2 - 2 3313 71
GenBank mRNA AI188219.1 - 1 - 1 71
AK125946.1 - 2 - 2 3313 71
AK126068.1 - 1 - 1 991 71
BC029521.1 - 1 - 1 1009 71
BC050288.1 - 3 - 3 2416 71

View Full Product Details