The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002022.2 | NP_002013.1 | 1 - 2 | 238 | 124 |
XM_005245045.3 | XP_005245102.1 | 1 - 2 | 289 | 124 | |
XM_005245046.3 | XP_005245103.1 | 1 - 2 | 290 | 124 | |
XM_005245048.3 | XP_005245105.1 | 1 - 2 | 290 | 124 | |
XM_006711243.3 | XP_006711306.1 | 1 - 2 | 290 | 124 | |
XM_011509347.2 | XP_011507649.1 | 1 - 2 | 290 | 124 | |
XM_011509348.2 | XP_011507650.1 | 1 - 2 | 289 | 124 | |
XM_011509349.2 | XP_011507651.1 | 1 - 2 | 288 | 124 | |
GenBank mRNA | BC002780.2 | - | 1 - 2 | 187 | 124 |
BX495448.1 | - | 1 - 2 | 240 | 124 | |
Z11737.1 | - | 1 - 2 | 74 | 124 |