Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPAR5
Entrez Gene ID
57121
Gene Name
lysophosphatidic acid receptor 5
Gene Aliases
GPR92, GPR93, KPG_010, LPA5
Chromosome Location
Chr.12: 6618835 - 6636131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142961.1 NP_001136433.1 1 - 2 64 70
GenBank mRNA DC328599.1 - 1 - 2 65 70

View Full Product Details