The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012382.2 | NP_036514.1 | 2 - 3 | 375 | 125 |
XM_011514003.2 | XP_011512305.1 | 2 - 3 | 1500 | 125 | |
GenBank mRNA | AF023244.1 | - | 2 - 3 | 321 | 125 |
AK312560.1 | - | 2 - 3 | 349 | 125 | |
BC015701.1 | - | 2 - 3 | 345 | 125 | |
BC036536.1 | - | 2 - 3 | 324 | 125 | |
DA518274.1 | - | 2 - 3 | 375 | 125 |