The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032367.1 | NP_001027539.1 | 2 - 3 | 705 | 78 |
NM_003710.3 | NP_003701.1 | 3 - 4 | 799 | 78 | |
NM_181642.2 | NP_857593.1 | 3 - 4 | 799 | 78 | |
XM_006720657.1 | XP_006720720.1 | 3 - 4 | 762 | 78 | |
XM_011521957.1 | XP_011520259.1 | 3 - 4 | 829 | 78 | |
XM_011521958.1 | XP_011520260.1 | 3 - 4 | 829 | 78 | |
GenBank mRNA | AB000095.1 | - | 3 - 4 | 770 | 78 |
AK300195.1 | - | 4 - 5 | 700 | 78 | |
AK303899.1 | - | 3 - 4 | 545 | 78 | |
AK314825.1 | - | 3 - 4 | 830 | 78 | |
AY296715.1 | - | 3 - 4 | 682 | 78 | |
AY358969.1 | - | 3 - 4 | 782 | 78 | |
BC004140.1 | - | 2 - 3 | 647 | 78 | |
BC018702.1 | - | 3 - 4 | 791 | 78 | |
BT007425.1 | - | 2 - 3 | 595 | 78 |