The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323505.1 | NP_001310434.1 | 11 - 12 | 1868 | 83 |
NM_001323506.1 | NP_001310435.1 | 11 - 12 | 1871 | 83 | |
NM_003999.2 | NP_003990.1 | 11 - 12 | 1988 | 83 | |
XM_005248384.1 | XP_005248441.1 | 11 - 12 | 1978 | 83 | |
XM_011514161.2 | XP_011512463.1 | 11 - 12 | 1978 | 83 | |
XM_017010019.1 | XP_016865508.1 | 11 - 12 | 1975 | 83 | |
XM_017010020.1 | XP_016865509.1 | 5 - 6 | 678 | 83 | |
GenBank mRNA | AK304016.1 | - | 6 - 7 | 996 | 83 |
BC125209.1 | - | 11 - 12 | 1758 | 83 | |
BC125210.1 | - | 11 - 12 | 1758 | 83 | |
U60805.1 | - | 11 - 12 | 1953 | 83 |