Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTP15
Entrez Gene ID
84135
Gene Name
UTP15, small subunit processome component
Gene Aliases
NET21
Chromosome Location
Chr.5: 73565741 - 73583377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284430.1 NP_001271359.1 11 - 12 1480 132
NM_001284431.1 NP_001271360.1 10 - 11 1364 132
NM_032175.3 NP_115551.2 11 - 12 1537 132
XM_011543680.2 XP_011541982.1 11 - 12 1798 132
GenBank mRNA AK022849.1 - 5 - 6 601 132
AK074217.1 - 11 - 12 1505 132
AK300525.1 - 10 - 11 1362 132
AK302021.1 - 11 - 12 1448 132
AL831972.1 - 11 - 12 1523 132
BC013064.1 - 6 - 7 714 132
CR457295.1 - 5 - 6 583 132

View Full Product Details