The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018429.2 | NP_060899.2 | 35 - 36 | 7462 | 117 |
XM_011543511.2 | XP_011541813.1 | 35 - 36 | 7434 | 117 | |
XM_011543512.2 | XP_011541814.1 | 32 - 33 | 6922 | 117 | |
XM_017009630.1 | XP_016865119.1 | 35 - 36 | 7441 | 117 | |
XM_017009631.1 | XP_016865120.1 | 35 - 36 | 7441 | 117 | |
XM_017009632.1 | XP_016865121.1 | 35 - 36 | 7440 | 117 | |
XM_017009633.1 | XP_016865122.1 | 34 - 35 | 7253 | 117 | |
XM_017009634.1 | XP_016865123.1 | 33 - 34 | 7108 | 117 | |
GenBank mRNA | AB051476.3 | - | 35 - 36 | 7209 | 117 |
BC146792.1 | - | 35 - 36 | 7209 | 117 | |
BC161615.1 | - | 35 - 36 | 7191 | 117 | |
BX641011.1 | - | 2 - 3 | 233 | 117 | |
CR749229.1 | - | 10 - 11 | 1265 | 117 | |
CR749289.1 | - | 13 - 14 | 1831 | 117 |