Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL11
Entrez Gene ID
3589
Gene Name
interleukin 11
Gene Aliases
AGIF, IL-11
Chromosome Location
Chr.19: 55364382 - 55370463 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000641.3 NP_000632.1 1 - 2 154 107
GenBank mRNA AK290572.1 - 1 - 2 154 107
AK315294.1 - 1 - 2 154 107
BC012506.1 - 1 - 2 137 107
M57765.1 - 1 - 2 137 107

View Full Product Details