Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL21
Entrez Gene ID
59067
Gene Name
interleukin 21
Gene Aliases
CVID11, IL-21, Za11
Chromosome Location
Chr.4: 122612628 - 122621057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207006.2 NP_001193935.1 2 - 3 248 102
NM_021803.3 NP_068575.1 2 - 3 248 102
GenBank mRNA AF254069.1 - 2 - 3 248 102
BC066258.1 - 2 - 3 247 102
BC066259.1 - 2 - 3 247 102
BC066260.1 - 2 - 3 247 102
BC066261.1 - 2 - 3 247 102
BC066262.1 - 2 - 3 247 102
BC069124.1 - 2 - 3 242 102
CD559460.2 - 3 - 4 102
CD559613.2 - 2 - 3 280 102
DQ645417.1 - 2 - 3 202 102

View Full Product Details