The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163257.1 | NP_001156729.1 | 12 - 13 | 2475 | 58 |
NM_005393.2 | NP_005384.2 | 11 - 12 | 2309 | 58 | |
GenBank mRNA | AB033032.1 | - | 11 - 12 | 2249 | 58 |
AF149019.1 | - | 11 - 12 | 2249 | 58 | |
AK292535.1 | - | 11 - 12 | 2309 | 58 | |
AK295762.1 | - | 11 - 12 | 2329 | 58 | |
AK295849.1 | - | 8 - 9 | 999 | 58 | |
AK295894.1 | - | 10 - 11 | 1193 | 58 | |
AK304705.1 | - | 11 - 12 | 1374 | 58 | |
AK307894.1 | - | 12 - 13 | 2452 | 58 |