Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLXNB3
Entrez Gene ID
5365
Gene Name
plexin B3
Gene Aliases
PLEXB3, PLEXR, PLXN6
Chromosome Location
Chr.X: 153764196 - 153779346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163257.1 NP_001156729.1 18 - 19 3246 77
NM_005393.2 NP_005384.2 17 - 18 3080 77
GenBank mRNA AB033032.1 - 17 - 18 3020 77
AF149019.1 - 17 - 18 3020 77
AK292535.1 - 17 - 18 3080 77
AK295762.1 - 17 - 18 3100 77
AK295849.1 - 14 - 15 1770 77
AK295894.1 - 16 - 17 1964 77
AK307894.1 - 18 - 19 3223 77

View Full Product Details