Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLXNB3
Entrez Gene ID
5365
Gene Name
plexin B3
Gene Aliases
PLEXB3, PLEXR, PLXN6
Chromosome Location
Chr.X: 153764196 - 153779346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163257.1 NP_001156729.1 6 - 7 1737 87
NM_005393.2 NP_005384.2 5 - 6 1571 87
GenBank mRNA AB033032.1 - 5 - 6 1511 87
AF149019.1 - 5 - 6 1511 87
AK292535.1 - 5 - 6 1571 87
AK295762.1 - 5 - 6 1591 87
AK295894.1 - 4 - 5 455 87
AK304705.1 - 5 - 6 636 87
AK307894.1 - 6 - 7 1714 87

View Full Product Details