Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCF4
Entrez Gene ID
4689
Gene Name
neutrophil cytosolic factor 4
Gene Aliases
CGD3, NCF, P40PHOX, SH3PXD4
Chromosome Location
Chr.22: 36860988 - 36878017 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000631.4 NP_000622.2 5 - 6 657 117
NM_013416.3 NP_038202.2 5 - 6 657 117
XM_017028808.1 XP_016884297.1 5 - 6 1021 117
GenBank mRNA AB025219.1 - 5 - 6 509 117
AB025220.1 - 5 - 6 509 117
AK223324.1 - 5 - 6 628 117
AK290924.1 - 5 - 6 579 117
BC002798.1 - 5 - 6 657 117
BT007346.1 - 5 - 6 473 117
BX355636.2 - 5 - 6 633 117
CR456528.1 - 5 - 6 554 117
CR542078.1 - 5 - 6 473 117
X77094.1 - 5 - 6 603 117

View Full Product Details