The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261824.1 | NP_001248753.1 | 4 - 5 | 561 | 79 |
NM_001320795.1 | NP_001307724.1 | 4 - 5 | 764 | 79 | |
NM_003720.3 | NP_003711.1 | 4 - 5 | 576 | 79 | |
NR_049728.1 | 3 - 4 | 469 | 79 | ||
GenBank mRNA | AF417108.1 | - | 3 - 4 | 340 | 79 |
AJ006291.1 | - | 4 - 5 | 510 | 79 | |
AK001455.1 | - | 4 - 5 | 538 | 79 | |
AK308614.1 | - | 3 - 4 | 447 | 79 | |
BC003619.2 | - | 4 - 5 | 493 | 79 | |
BC010424.1 | - | 4 - 5 | 485 | 79 | |
BC011755.2 | - | 4 - 5 | 493 | 79 | |
BC012809.2 | - | 4 - 5 | 493 | 79 | |
BM460414.1 | - | 4 - 5 | 469 | 79 | |
BM462074.1 | - | 4 - 5 | 493 | 79 | |
BQ008489.1 | - | 3 - 4 | 79 | ||
BU543452.1 | - | 4 - 5 | 527 | 79 | |
CR541821.1 | - | 4 - 5 | 447 | 79 | |
CR541852.1 | - | 4 - 5 | 447 | 79 |