Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCALD
Entrez Gene ID
83988
Gene Name
neurocalcin delta
Gene Aliases
-
Chromosome Location
Chr.8: 101686542 - 102124907 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032041.2 NP_114430.2 1 - 2 347 74
GenBank mRNA AB209015.1 - 1 - 2 87 74
AK055073.1 - 1 - 2 105 74
AK314523.1 - 1 - 2 106 74
AL136612.1 - 1 - 2 98 74
DA211806.1 - 1 - 2 347 74

View Full Product Details