The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182924.3 | NP_891554.1 | 10 - 11 | 2270 | 80 |
XM_011515520.2 | XP_011513822.1 | 10 - 11 | 2294 | 80 | |
XM_011515522.2 | XP_011513824.1 | 10 - 11 | 2294 | 80 | |
XM_011515523.2 | XP_011513825.1 | 10 - 11 | 2300 | 80 | |
XM_011515524.2 | XP_011513826.1 | 4 - 5 | 494 | 80 | |
GenBank mRNA | AK074068.1 | - | 9 - 10 | 3671 | 80 |
AK093249.1 | - | 1 - 2 | 1035 | 80 | |
AK126808.1 | - | 3 - 4 | 1252 | 80 | |
AL833704.2 | - | 9 - 10 | 2224 | 80 | |
BC037988.1 | - | 10 - 11 | 2257 | 80 |