Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EEPD1
Entrez Gene ID
80820
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
HSPC107
Chromosome Location
Chr.7: 36153149 - 36301543 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030636.2 NP_085139.2 7 - 8 2147 85
GenBank mRNA AB051493.1 - 7 - 8 1888 85
AK027386.1 - 7 - 8 2119 85
BC065518.1 - 8 - 9 2047 85

View Full Product Details