The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286791.1 | NP_001273720.1 | 9 - 10 | 1367 | 79 |
NM_025087.2 | NP_079363.2 | 9 - 10 | 1448 | 79 | |
XM_011513755.1 | XP_011512057.1 | 9 - 10 | 1449 | 79 | |
XM_011513756.2 | XP_011512058.1 | 9 - 10 | 1480 | 79 | |
XM_011513757.2 | XP_011512059.1 | 9 - 10 | 1506 | 79 | |
XM_011513758.1 | XP_011512060.1 | 8 - 9 | 1191 | 79 | |
XM_011513759.1 | XP_011512061.1 | 6 - 7 | 845 | 79 | |
GenBank mRNA | AK025164.1 | - | 9 - 10 | 1448 | 79 |
AK300495.1 | - | 9 - 10 | 1370 | 79 | |
AK316307.1 | - | 9 - 10 | 1340 | 79 | |
BC137387.1 | - | 9 - 10 | 1431 | 79 | |
BC137388.1 | - | 9 - 10 | 1431 | 79 |