Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8orf74
Entrez Gene ID
203076
Gene Name
chromosome 8 open reading frame 74
Gene Aliases
-
Chromosome Location
Chr.8: 10672557 - 10700616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040032.1 NP_001035121.1 2 - 3 273 58
XM_011543816.2 XP_011542118.1 2 - 3 374 58
GenBank mRNA BC065837.1 - 2 - 3 273 58
BC132852.1 - 2 - 3 273 58
BC132854.1 - 2 - 3 273 58

View Full Product Details