The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320289.1 | NP_001307218.1 | 3 - 4 | 573 | 72 |
NM_001320290.1 | NP_001307219.1 | 3 - 4 | 310 | 72 | |
NM_178033.1 | NP_828847.1 | 3 - 4 | 607 | 72 | |
XM_017000973.1 | XP_016856462.1 | 3 - 4 | 638 | 72 | |
GenBank mRNA | AK091806.1 | - | 3 - 4 | 311 | 72 |
AK098065.1 | - | 3 - 4 | 600 | 72 | |
AK131355.1 | - | 3 - 4 | 573 | 72 | |
AM040940.1 | - | 3 - 4 | 444 | 72 | |
AY358537.1 | - | 3 - 4 | 590 | 72 | |
BC028102.1 | - | 3 - 4 | 607 | 72 |