The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206855.1 | NP_001193784.1 | 9 - 10 | 1160 | 59 |
NM_003227.3 | NP_003218.2 | 12 - 13 | 1580 | 59 | |
XM_005250553.4 | XP_005250610.1 | 13 - 14 | 1886 | 59 | |
XM_017012573.1 | XP_016868062.1 | 13 - 14 | 1688 | 59 | |
GenBank mRNA | AF067864.1 | - | 12 - 13 | 1566 | 59 |
AK000421.1 | - | 8 - 9 | 892 | 59 | |
BC142630.1 | - | 12 - 13 | 1566 | 59 | |
BC150192.1 | - | 9 - 10 | 1374 | 59 | |
BI917390.1 | - | 5 - 6 | 612 | 59 |