Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
Chromosome Location
Chr.7: 100620416 - 100642780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017012573.1 XP_016868062.1 1 - 2 86 95
GenBank mRNA AK022002.1 - 1 - 2 95 95

View Full Product Details