The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014920.3 | NP_055735.1 | 7 - 8 | 1044 | 113 |
NM_016513.4 | NP_057597.2 | 8 - 9 | 1155 | 113 | |
XM_011514419.1 | XP_011512721.1 | 7 - 8 | 1044 | 113 | |
XM_011514420.2 | XP_011512722.1 | 8 - 9 | 1186 | 113 | |
XM_011514421.1 | XP_011512723.1 | 8 - 9 | 1125 | 113 | |
XM_017010485.1 | XP_016865974.1 | 8 - 9 | 1418 | 113 | |
XM_017010486.1 | XP_016865975.1 | 9 - 10 | 1297 | 113 | |
XM_017010487.1 | XP_016865976.1 | 7 - 8 | 963 | 113 | |
XM_017010488.1 | XP_016865977.1 | 8 - 9 | 1845 | 113 | |
XM_017010489.1 | XP_016865978.1 | 9 - 10 | 1360 | 113 | |
XM_017010490.1 | XP_016865979.1 | 8 - 9 | 1230 | 113 | |
XM_017010491.1 | XP_016865980.1 | 8 - 9 | 1191 | 113 | |
XM_017010492.1 | XP_016865981.1 | 7 - 8 | 1235 | 113 | |
GenBank mRNA | AB023153.1 | - | 7 - 8 | 931 | 113 |
AF152469.1 | - | 7 - 8 | 1012 | 113 | |
AF225919.1 | - | 7 - 8 | 874 | 113 | |
BC035807.1 | - | 7 - 8 | 1076 | 113 | |
BC136420.1 | - | 6 - 7 | 705 | 113 | |
BC136421.1 | - | 6 - 7 | 705 | 113 | |
BC152464.1 | - | 7 - 8 | 931 | 113 |