The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014979.2 | NP_001014979.2 | 2 - 3 | 734 | 85 |
NM_001195620.1 | NP_001182549.1 | 2 - 3 | 734 | 85 | |
XM_011545976.1 | XP_011544278.1 | 2 - 3 | 767 | 85 | |
XM_011545978.1 | XP_011544280.1 | 2 - 3 | 767 | 85 | |
XM_017023852.1 | XP_016879341.1 | 2 - 3 | 767 | 85 | |
XM_017023854.1 | XP_016879343.1 | 2 - 3 | 767 | 85 | |
XM_017023855.1 | XP_016879344.1 | 2 - 3 | 767 | 85 | |
XM_017023856.1 | XP_016879345.1 | 2 - 3 | 288 | 191 | |
XM_017023858.1 | XP_016879347.1 | 2 - 3 | 767 | 85 | |
XM_017023859.1 | XP_016879348.1 | 2 - 3 | 767 | 85 | |
XM_017023861.1 | XP_016879350.1 | 2 - 3 | 767 | 85 | |
GenBank mRNA | BC073881.1 | - | 2 - 3 | 568 | 85 |
BC128128.1 | - | 2 - 3 | 238 | 85 | |
BC128129.1 | - | 2 - 3 | 238 | 85 | |
BI829598.1 | - | 2 - 3 | 730 | 84 |