Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRGAP3
Entrez Gene ID
9901
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
Gene Aliases
ARHGAP14, MEGAP, SRGAP2, WRP
Chromosome Location
Chr.3: 8980591 - 9362979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033117.2 NP_001028289.1 11 - 12 2172 94
XM_017007575.1 XP_016863064.1 11 - 12 2510 94
XM_017007580.1 XP_016863069.1 13 - 14 1739 94
GenBank mRNA AB007871.2 - 11 - 12 1396 94
AF464189.1 - 11 - 12 1700 94

View Full Product Details