Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRGAP3
Entrez Gene ID
9901
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
Gene Aliases
ARHGAP14, MEGAP, SRGAP2, WRP
Chromosome Location
Chr.3: 8980591 - 9362979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033117.2 NP_001028289.1 7 - 8 1760 80
NM_014850.3 NP_055665.1 7 - 8 1760 80
XM_011534295.1 XP_011532597.1 7 - 8 1760 80
XM_011534296.1 XP_011532598.1 8 - 9 1877 80
XM_011534301.2 XP_011532603.1 4 - 5 3589 80
XM_011534303.1 XP_011532605.1 4 - 5 603 80
XM_017007574.1 XP_016863063.1 7 - 8 2098 80
XM_017007575.1 XP_016863064.1 7 - 8 2098 80
XM_017007576.1 XP_016863065.1 9 - 10 1267 80
XM_017007577.1 XP_016863066.1 9 - 10 1206 80
XM_017007578.1 XP_016863067.1 7 - 8 2126 80
XM_017007579.1 XP_016863068.1 7 - 8 2094 80
XM_017007580.1 XP_016863069.1 9 - 10 1327 80
GenBank mRNA AB007871.2 - 7 - 8 984 80
AB032982.1 - 5 - 6 635 80
AF427144.1 - 7 - 8 1128 80
AF464189.1 - 7 - 8 1288 80
BC039300.1 - 7 - 8 1095 80

View Full Product Details