Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5C2
Entrez Gene ID
22978
Gene Name
5'-nucleotidase, cytosolic II
Gene Aliases
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location
Chr.10: 103088017 - 103193306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134373.2 NP_001127845.1 17 - 18 1543 99
NM_012229.4 NP_036361.1 18 - 19 1641 99
XM_005269632.4 XP_005269689.1 18 - 19 1552 99
XM_005269633.4 XP_005269690.1 19 - 20 1645 99
XM_005269634.4 XP_005269691.1 19 - 20 1677 99
XM_005269635.4 XP_005269692.1 18 - 19 1867 99
XM_005269636.4 XP_005269693.1 18 - 19 1653 99
XM_005269637.4 XP_005269694.1 17 - 18 2371 99
XM_005269638.4 XP_005269695.1 17 - 18 3672 99
XM_005269639.4 XP_005269696.1 16 - 17 1396 99
XM_005269640.4 XP_005269697.1 19 - 20 1668 99
XM_005269641.4 XP_005269698.1 18 - 19 1570 99
XM_005269642.4 XP_005269699.1 17 - 18 1495 99
XM_005269643.4 XP_005269700.1 16 - 17 1435 99
XM_005269644.4 XP_005269701.1 20 - 21 1692 99
XM_005269645.4 XP_005269702.1 19 - 20 1594 99
XM_005269646.4 XP_005269703.1 18 - 19 1520 99
XM_006717721.3 XP_006717784.1 18 - 19 1594 99
XM_006717723.3 XP_006717786.1 16 - 17 1391 99
XM_011539537.2 XP_011537839.1 18 - 19 1506 99
XM_017015947.1 XP_016871436.1 16 - 17 1421 99
XM_017015948.1 XP_016871437.1 19 - 20 1777 99
XM_017015949.1 XP_016871438.1 19 - 20 1762 99
XM_017015950.1 XP_016871439.1 17 - 18 1478 99
XM_017015951.1 XP_016871440.1 16 - 17 1404 99
XM_017015952.1 XP_016871441.1 18 - 19 1701 99
XM_017015953.1 XP_016871442.1 19 - 20 2096 99
XM_017015954.1 XP_016871443.1 18 - 19 2035 99
XM_017015955.1 XP_016871444.1 18 - 19 1722 99
XM_017015956.1 XP_016871445.1 18 - 19 1928 99
XM_017015957.1 XP_016871446.1 20 - 21 1786 99
XM_017015958.1 XP_016871447.1 21 - 22 1905 99
XM_017015959.1 XP_016871448.1 17 - 18 1496 99
XM_017015960.1 XP_016871449.1 18 - 19 1459 99
XM_017015961.1 XP_016871450.1 18 - 19 1854 99
XM_017015962.1 XP_016871451.1 18 - 19 1688 99
XM_017015963.1 XP_016871452.1 18 - 19 1792 99
XM_017015964.1 XP_016871453.1 18 - 19 1771 99
XM_017015965.1 XP_016871454.1 17 - 18 1648 99
XM_017015966.1 XP_016871455.1 17 - 18 1648 99
XM_017015967.1 XP_016871456.1 17 - 18 1583 99
XM_017015968.1 XP_016871457.1 18 - 19 1576 99
XM_017015969.1 XP_016871458.1 18 - 19 1670 99
XM_017015970.1 XP_016871459.1 16 - 17 2313 99
XM_017015971.1 XP_016871460.1 15 - 16 1357 99
XM_017015972.1 XP_016871461.1 19 - 20 1611 99
XM_017015973.1 XP_016871462.1 20 - 21 1635 99
XM_017015974.1 XP_016871463.1 19 - 20 1635 99
XM_017015975.1 XP_016871464.1 17 - 18 1476 99
XM_017015976.1 XP_016871465.1 17 - 18 1482 99
GenBank mRNA AK127670.1 - 18 - 19 1632 99
AK291667.1 - 17 - 18 1509 99
AK295593.1 - 16 - 17 1685 99
AK312517.1 - 17 - 18 1509 99
BC001595.1 - 17 - 18 1592 99
BX648078.1 - 17 - 18 5543 99
D38524.1 - 17 - 18 1531 99

View Full Product Details