Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5C2
Entrez Gene ID
22978
Gene Name
5'-nucleotidase, cytosolic II
Gene Aliases
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location
Chr.10: 103088017 - 103193306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134373.2 NP_001127845.1 2 - 3 196 147
NM_012229.4 NP_036361.1 3 - 4 294 147
XM_005269632.4 XP_005269689.1 2 - 3 181 147
XM_005269633.4 XP_005269690.1 3 - 4 274 147
XM_005269634.4 XP_005269691.1 3 - 4 306 147
XM_005269635.4 XP_005269692.1 2 - 3 496 147
XM_005269636.4 XP_005269693.1 3 - 4 306 147
XM_005269640.4 XP_005269697.1 3 - 4 260 208
XM_005269641.4 XP_005269698.1 2 - 3 162 208
XM_005269644.4 XP_005269701.1 3 - 4 260 208
XM_005269645.4 XP_005269702.1 2 - 3 162 208
XM_011539537.2 XP_011537839.1 3 - 4 306 147
XM_017015948.1 XP_016871437.1 3 - 4 306 271
XM_017015949.1 XP_016871438.1 2 - 3 162 208
XM_017015950.1 XP_016871439.1 2 - 3 162 208
XM_017015952.1 XP_016871441.1 2 - 3 162 147
XM_017015953.1 XP_016871442.1 2 - 3 496 208
XM_017015954.1 XP_016871443.1 2 - 3 496 147
XM_017015955.1 XP_016871444.1 2 - 3 162 147
XM_017015956.1 XP_016871445.1 2 - 3 496 147
XM_017015957.1 XP_016871446.1 2 - 3 162 208
XM_017015958.1 XP_016871447.1 3 - 4 260 208
XM_017015968.1 XP_016871457.1 3 - 4 260 208
XM_017015969.1 XP_016871458.1 2 - 3 162 208
XM_017015972.1 XP_016871461.1 2 - 3 162 208
XM_017015973.1 XP_016871462.1 2 - 3 162 208
XM_017015976.1 XP_016871465.1 1 - 2 125 208
GenBank mRNA AK291667.1 - 2 - 3 162 147
AK312517.1 - 2 - 3 162 147
BC001595.1 - 2 - 3 245 147
BX648078.1 - 2 - 3 242 208
D38524.1 - 2 - 3 184 147
DA879821.1 - 3 - 4 263 147
DB169780.1 - 2 - 3 198 147

View Full Product Details