Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
Chromosome Location
Chr.14: 77274956 - 77320885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013382.5 NP_037514.2 13 - 14 1685 84
XM_011536675.2 XP_011534977.1 13 - 14 1685 84
XM_011536676.2 XP_011534978.1 12 - 13 1598 84
XM_011536677.2 XP_011534979.1 9 - 10 1227 84
XM_011536679.2 XP_011534981.1 9 - 10 887 84
XM_017021244.1 XP_016876733.1 12 - 13 1519 84
XM_017021245.1 XP_016876734.1 9 - 10 973 84
GenBank mRNA AB209114.1 - 13 - 14 1507 84
AF105020.1 - 13 - 14 1688 84
AK025962.1 - 3 - 4 304 84
AY090480.1 - 13 - 14 1688 84
BC031651.1 - 13 - 14 1575 84
BE797202.1 - - 91 84
CR627447.1 - 13 - 14 1255 84

View Full Product Details