Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
Chromosome Location
Chr.14: 77274956 - 77320885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013382.5 NP_037514.2 7 - 8 1124 71
XM_011536675.2 XP_011534977.1 7 - 8 1124 71
XM_011536676.2 XP_011534978.1 6 - 7 1037 71
XM_011536679.2 XP_011534981.1 3 - 4 326 71
XM_017021244.1 XP_016876733.1 6 - 7 958 71
XM_017021245.1 XP_016876734.1 3 - 4 412 71
GenBank mRNA AB209114.1 - 7 - 8 946 71
AF105020.1 - 7 - 8 1127 71
AK023804.1 - 1 - 2 1715 71
AY090480.1 - 7 - 8 1127 71
BC031651.1 - 7 - 8 1014 71
BM127609.1 - 1 - 2 48 71
CR627447.1 - 7 - 8 694 71

View Full Product Details