The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000293.2 | NP_000284.1 | 5 - 6 | 563 | 87 |
NM_001031835.2 | NP_001027005.1 | 6 - 7 | 675 | 87 | |
XM_005255983.4 | XP_005256040.1 | 5 - 6 | 526 | 87 | |
XM_005255984.4 | XP_005256041.1 | 6 - 7 | 826 | 87 | |
XM_017023283.1 | XP_016878772.1 | 6 - 7 | 648 | 87 | |
XM_017023284.1 | XP_016878773.1 | 5 - 6 | 529 | 87 | |
GenBank mRNA | AK309226.1 | - | 5 - 6 | 526 | 87 |
BC033657.1 | - | 6 - 7 | 654 | 87 | |
CR936710.1 | - | 6 - 7 | 628 | 87 | |
X84908.1 | - | 5 - 6 | 535 | 87 |