Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHKB
Entrez Gene ID
5257
Gene Name
phosphorylase kinase regulatory subunit beta
Gene Aliases
-
Chromosome Location
Chr.16: 47461131 - 47701523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000293.2 NP_000284.1 8 - 9 826 105
NM_001031835.2 NP_001027005.1 9 - 10 938 105
XM_005255983.4 XP_005256040.1 8 - 9 789 105
XM_005255984.4 XP_005256041.1 9 - 10 1089 105
XM_017023282.1 XP_016878771.1 2 - 3 83 105
XM_017023283.1 XP_016878772.1 9 - 10 911 105
XM_017023284.1 XP_016878773.1 8 - 9 792 105
GenBank mRNA AK309226.1 - 8 - 9 789 105
BC033657.1 - 9 - 10 917 105
CR936710.1 - 9 - 10 972 105
X84908.1 - 8 - 9 798 105

View Full Product Details