The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000922.3 | NP_000913.2 | 6 - 7 | 2087 | 131 |
XM_006718249.3 | XP_006718312.1 | 6 - 7 | 2119 | 131 | |
XM_017017911.1 | XP_016873400.1 | 6 - 7 | 2119 | 131 | |
XM_017017912.1 | XP_016873401.1 | 6 - 7 | 2119 | 131 | |
GenBank mRNA | AY459346.1 | - | 6 - 7 | 2025 | 131 |
BC136565.1 | - | 6 - 7 | 1980 | 131 | |
BC136566.1 | - | 6 - 7 | 1980 | 131 | |
BC144248.1 | - | 6 - 7 | 1827 | 131 | |
BC150307.1 | - | 6 - 7 | 2087 | 131 | |
X95520.1 | - | 6 - 7 | 1734 | 131 |