The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318976.1 | NP_001305905.1 | 10 - 11 | 1208 | 123 |
NM_001318977.1 | NP_001305906.1 | 9 - 10 | 1078 | 123 | |
NM_005680.2 | NP_005671.2 | 10 - 11 | 1232 | 123 | |
XM_011510410.2 | XP_011508712.1 | 10 - 11 | 1173 | 123 | |
XM_011510411.1 | XP_011508713.1 | 11 - 12 | 1281 | 123 | |
GenBank mRNA | AK026521.1 | - | 9 - 10 | 995 | 123 |
AK295402.1 | - | 10 - 11 | 1127 | 123 | |
AK308070.1 | - | 10 - 11 | 1181 | 123 | |
AK309751.1 | - | 10 - 11 | 1215 | 123 | |
AK315098.1 | - | 10 - 11 | 1164 | 123 | |
BC018137.2 | - | 10 - 11 | 1231 | 123 | |
L39061.1 | - | 10 - 11 | 1234 | 123 |